Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000411826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000411826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000411826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000411826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000411826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000441954.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
ENST00000441954.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
NM_153612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
NM_153612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
NM_153612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
NM_153612.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012932.hg.1 |
NM_153612.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000312719.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000411826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000411826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000411826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000411826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000411826.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000441954.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
ENST00000441954.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
NM_153612.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
NM_153612.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
NM_153612.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
NM_153612.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012932.hg.1 |
NM_153612.3 |
Refseq |