Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000265814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000360348.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000396218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000422361.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000436581.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000517493.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000517792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000517919.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000518256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000518317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000518823.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000518832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000518844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000518954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000518992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000519061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000519422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000519577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000519847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000520172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000520428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000520556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000520583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000520724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000520974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000521054.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000521319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000521375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000521553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000521733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000521897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000522065.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000522467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000523168.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000523290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000523629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000524215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000613302.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000613886.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000614812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000615601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000617740.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198626.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198627.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198679.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_004349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_175634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_175635.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_175636.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000265814.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000360348.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000396218.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000422361.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000436581.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000517493.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000517792.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000517919.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000518256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000518317.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000518823.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000518832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000518844.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000518954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000518992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000519061.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000519422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000519577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000519847.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000520172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000520428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000520556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000520583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000520724.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000520974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000521054.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000521319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000521375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000521553.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000521733.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000521897.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000522065.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000522467.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000523168.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000523290.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000523629.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000524215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000613302.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000613886.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000614812.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000615601.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
ENST00000617740.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198625.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198626.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198627.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198630.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198631.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198633.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198634.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_001198679.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_004349.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_175634.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_175635.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
NM_175636.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800175469.hg.1 |
TC0800011086.hg.1 |
Clariom D TC |