Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006864.hg.1 |
ENST00000371939.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006864.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006864.hg.1 |
TC10000626.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000531458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000531458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000531458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000531458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
ENST00000531458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006865.hg.1 |
ENST00000371939.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006865.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006865.hg.1 |
TC10000626.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000156717.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156717.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156717.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156717.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156717.hg.1 |
TC1000008377.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000156719.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156719.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156719.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156719.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156719.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156719.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156719.hg.1 |
TC1000008377.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000371939.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000526923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000531458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000531458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000531458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000531458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
ENST00000531458.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008377.hg.1 |
NM_001010939.2 |
Refseq |