Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000517720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000517720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000517720.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000524341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000524341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000524341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000524341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000524341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
ENST00000524341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000517720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000517720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000517720.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000524341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000524341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000524341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000524341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000524341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
ENST00000524341.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011172.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176116.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176116.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176116.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176116.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176116.hg.1 |
TC0800011172.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800176117.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176117.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176117.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176117.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176117.hg.1 |
TC0800011172.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800176118.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176118.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176118.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176118.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176118.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176118.hg.1 |
ENST00000287025.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176118.hg.1 |
ENST00000522822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176118.hg.1 |
ENST00000523821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176118.hg.1 |
NM_001286643.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176118.hg.1 |
NM_015942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176118.hg.1 |
TC0800011172.hg.1 |
Clariom D TC |