Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021101.hg.1 |
TC03001225.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021101.hg.1 |
ENST00000295824.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021101.hg.1 |
ENST00000344838.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021101.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03021101.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000467602.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000467602.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000467602.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000467602.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207000.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207000.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207000.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207000.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207000.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207000.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207000.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207000.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207000.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300207000.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300207000.hg.1 |
TC0300013950.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000295824.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000389256.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000440022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000467602.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000467602.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000467602.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000467602.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
ENST00000498089.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013950.hg.1 |
NM_144715.3 |
Refseq |