Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029467.hg.1 |
TC03001631.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029467.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03029467.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000495753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000495753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000495753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000495753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
ENST00000495753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000478998.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000495753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000495753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000495753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000495753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
ENST00000495753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011941.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300188890.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188890.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300188890.hg.1 |
ENST00000273353.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300188890.hg.1 |
NM_014981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300188890.hg.1 |
TC0300011941.hg.1 |
Clariom D TC |