Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
TC02000432.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
ENST00000258104.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
ENST00000394120.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
ENST00000409366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
ENST00000409582.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
ENST00000409651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
ENST00000409744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
ENST00000409762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
ENST00000410020.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
ENST00000410041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
ENST00000413539.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
ENST00000429174.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
ENST00000479049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
NM_001130455.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
NM_001130976.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
NM_001130977.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
NM_001130978.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
NM_001130979.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
NM_001130980.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
NM_001130981.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
NM_001130982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
NM_001130983.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
NM_001130984.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
NM_001130985.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
NM_001130986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
NM_001130987.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02006636.hg.1 |
NM_003494.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000258104.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000394120.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000409366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000409582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000409651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000409744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000409762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000410020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000410041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000413539.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000429174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000479049.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130976.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130977.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_003494.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000258104.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000394120.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000409366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000409582.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000409651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000409744.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000409762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000410020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000410041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000413539.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000429174.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
ENST00000479049.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130455.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130976.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130977.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130978.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130979.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130980.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130981.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130983.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130984.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130985.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_001130987.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
NM_003494.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200155371.hg.1 |
TC0200008005.hg.1 |
Clariom D TC |