Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010057.hg.1 |
ENST00000366232.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000261854.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000558934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000559293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000559293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000559293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000559293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000559293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000559527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000559527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000559527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000559527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000559527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000559527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000559527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000559527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000560288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000560288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000560288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000560288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000560288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000560288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
ENST00000560288.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009396.hg.1 |
NM_032802.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010057.hg.1 |
ENST00000366232.3 |
Ensembl |