Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025191.hg.1 |
ENST00000320362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025191.hg.1 |
ENST00000402418.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025191.hg.1 |
ENST00000416858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025191.hg.1 |
ENST00000442286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025191.hg.1 |
NM_001126121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025191.hg.1 |
NM_001126122.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025191.hg.1 |
NM_021734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17025191.hg.1 |
TC17001876.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000320362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000375261.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000402418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000416858.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000442286.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000579207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000580151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000580994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000581988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000582778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000583332.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000584438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
NM_001126121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
NM_001126122.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
NM_021734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000320362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000375261.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000402418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000416858.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000442286.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000579207.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000580151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000580994.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000581988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000582778.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000583332.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
ENST00000584438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
NM_001126121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
NM_001126122.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
NM_021734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700195364.hg.1 |
TC1700011746.hg.1 |
Clariom D TC |