Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001303245.1 | NP_001290174.1 | 21 - 22 | 2267 | 139 | |
NM_001303246.1 | NP_001290175.1 | 22 - 23 | 2276 | 139 | ||
NM_006506.3 | NP_006497.2 | 21 - 22 | 2264 | 139 | ||
XM_011513059.2 | XP_011511361.1 | 22 - 23 | 3416 | 139 | ||
XM_017006968.1 | XP_016862457.1 | 22 - 23 | 2400 | 139 | ||
XM_017006969.1 | XP_016862458.1 | 22 - 23 | 2396 | 139 | ||
XM_017006972.1 | XP_016862461.1 | 20 - 21 | 2131 | 139 | ||
GenBank mRNA | AB209161.1 | - | 6 - 7 | 603 | 139 | |
AF115573.1 | - | 21 - 22 | 2227 | 139 | ||
AK292016.1 | - | 21 - 22 | 2265 | 139 | ||
D78155.1 | - | 21 - 22 | 2230 | 139 | ||
D82880.1 | - | 22 - 23 | 2239 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |