Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024163.hg.1 |
ENST00000343702.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024163.hg.1 |
ENST00000344911.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024163.hg.1 |
ENST00000538383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024163.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024163.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024163.hg.1 |
TC12001837.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000547980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000547980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000547980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000550074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000550074.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000552603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000552603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000552603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000552603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000547980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000547980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000547980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000550074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000550074.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000552603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000552603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000552603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000552603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011591.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189809.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189809.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189809.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189809.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189809.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189809.hg.1 |
ENST00000343702.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189809.hg.1 |
ENST00000344911.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189809.hg.1 |
ENST00000538383.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189809.hg.1 |
ENST00000553059.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200189809.hg.1 |
NM_021229.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200189809.hg.1 |
TC1200011591.hg.1 |
Clariom D TC |