Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000591584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000591584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000591584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000591584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
ENST00000591584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006500.hg.1 |
ENST00000253794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006500.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006500.hg.1 |
TC17000535.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006501.hg.1 |
ENST00000253794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006501.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17006501.hg.1 |
TC17000535.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000591584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000591584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000591584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000591584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
ENST00000591584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007914.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
ENST00000589219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
ENST00000589520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
ENST00000591584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
ENST00000589219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
ENST00000589520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
ENST00000591584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157699.hg.1 |
TC1700007914.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700157696.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157696.hg.1 |
ENST00000589219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157696.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157696.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157696.hg.1 |
ENST00000591584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157696.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157696.hg.1 |
ENST00000253794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157696.hg.1 |
ENST00000589219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157696.hg.1 |
ENST00000589577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157696.hg.1 |
ENST00000590339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157696.hg.1 |
ENST00000591584.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700157696.hg.1 |
NM_032353.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700157696.hg.1 |
TC1700007914.hg.1 |
Clariom D TC |