Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000588853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000588853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000588853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000592218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000592218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
ENST00000592218.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016535.hg.1 |
ENST00000359227.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016535.hg.1 |
ENST00000438662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016535.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016535.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19016535.hg.1 |
TC19001185.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000588853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000588853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000588853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000592218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000592218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
ENST00000592218.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011870.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204288.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204288.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204288.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204288.hg.1 |
NM_032281.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204288.hg.1 |
TC1900011870.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900204288.hg.1 |
ENST00000359227.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204288.hg.1 |
ENST00000438662.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204288.hg.1 |
NM_001420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204288.hg.1 |
NM_032281.2 |
Refseq |