Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019411.hg.1 |
TC09001542.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019411.hg.1 |
ENST00000223642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019411.hg.1 |
ENST00000430906.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019411.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000460578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000460578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000460578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000460578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000466280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000466280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000466280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000480188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000480188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000480188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000480188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000480188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000489802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000489802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
ENST00000489802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000460578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000460578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000460578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000460578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000466280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000466280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000466280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000480188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000480188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000480188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000480188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000480188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000489802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000489802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
ENST00000489802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011394.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177720.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177720.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177720.hg.1 |
ENST00000223642.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177720.hg.1 |
NM_001735.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177720.hg.1 |
TC0900011394.hg.1 |
Clariom D TC |