Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000474854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000474854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000474854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000474854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000474854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
ENST00000474854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148124.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148124.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148124.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148124.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148124.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148124.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600148124.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148124.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600148124.hg.1 |
TC0600007038.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000341776.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000397311.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000474854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000474854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000474854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000474854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000474854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
ENST00000474854.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_001267040.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007038.hg.1 |
NM_004973.3 |
Refseq |