Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026432.hg.1 |
TC05001912.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026432.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026432.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186324.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186324.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186324.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186324.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186324.hg.1 |
TC0500012394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000318315.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000510432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
ENST00000515291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012394.hg.1 |
NM_206966.2 |
Refseq |