Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168478.1 | NP_001161950.1 | 2 - 3 | 466 | 133 | |
NM_001168479.1 | NP_001161951.1 | 4 - 5 | 582 | 133 | ||
NM_001168480.1 | NP_001161952.1 | 4 - 5 | 576 | 133 | ||
GenBank mRNA | AK096692.1 | - | 4 - 5 | 576 | 133 | |
AL537480.3 | - | 2 - 3 | 386 | 133 | ||
BC050678.1 | - | 2 - 3 | 427 | 133 | ||
BI765968.1 | - | 4 - 5 | 534 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |