Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000475366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000475366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000475366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000475366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000475366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000491821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000491821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000491821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000495671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000496559.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000497721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000497721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000497721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000497721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000497721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000497721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000497721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000498287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000498287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000498287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000498287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000498287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000498287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
ENST00000498287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011423.hg.1 |
NM_015541.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185661.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185661.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185661.hg.1 |
ENST00000273261.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185661.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185661.hg.1 |
TC0300011423.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300185662.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185662.hg.1 |
ENST00000383703.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185662.hg.1 |
TC0300011423.hg.1 |
Clariom D TC |