Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000458235.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000526008.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000527670.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000528705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
ENST00000534444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009991.hg.1 |
NM_000215.3 |
Refseq |