Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000479456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000479456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000479456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000479456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000488084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000488084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000488084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000488084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000495687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000495687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000495687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000495687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000495687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
ENST00000495687.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000479456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000479456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000479456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000479456.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000488084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000488084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000488084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000488084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000495687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000495687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000495687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000495687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000495687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
ENST00000495687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013515.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198813.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198813.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198813.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198813.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198813.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198813.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198813.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198813.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198813.hg.1 |
TC0200013515.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200198814.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198814.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198814.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198814.hg.1 |
ENST00000495687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198814.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198814.hg.1 |
ENST00000337288.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198814.hg.1 |
ENST00000443962.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198814.hg.1 |
ENST00000462501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198814.hg.1 |
ENST00000495687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198814.hg.1 |
NM_020151.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198814.hg.1 |
TC0200013515.hg.1 |
Clariom D TC |