Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021463.hg.1 |
TC07001368.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021463.hg.1 |
ENST00000046087.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021463.hg.1 |
ENST00000419417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021463.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021463.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021468.hg.1 |
TC07001368.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021468.hg.1 |
ENST00000046087.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021468.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021468.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021465.hg.1 |
TC07001368.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021465.hg.1 |
ENST00000046087.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021465.hg.1 |
ENST00000419417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021465.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021465.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000450231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000450231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000450231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000450231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000450231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
ENST00000450231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200674.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200674.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200674.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200674.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200674.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200674.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200674.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200674.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200674.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200674.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200674.hg.1 |
TC0700013554.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000450231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000450231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000450231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000450231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000450231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
ENST00000450231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013554.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200671.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200671.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200671.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200671.hg.1 |
ENST00000450231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200671.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200671.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200671.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200671.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200671.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200671.hg.1 |
ENST00000450231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200671.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200671.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200671.hg.1 |
TC0700013554.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700200673.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200673.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200673.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200673.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200673.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200673.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200673.hg.1 |
ENST00000413331.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200673.hg.1 |
ENST00000419417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200673.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200673.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200673.hg.1 |
TC0700013554.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700200677.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200677.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200677.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200677.hg.1 |
ENST00000046087.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700200677.hg.1 |
NM_001159878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200677.hg.1 |
NM_007009.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700200677.hg.1 |
TC0700013554.hg.1 |
Clariom D TC |