Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPRIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319842.1 | NP_001306771.1 | 4 - 5 | 1558 | 88 | |
XM_017019874.1 | XP_016875363.1 | 5 - 6 | 776 | 88 | ||
XM_017019876.1 | XP_016875365.1 | 3 - 4 | 387 | 88 | ||
XM_017019877.1 | XP_016875366.1 | 3 - 4 | 387 | 88 | ||
XM_017019878.1 | XP_016875367.1 | 3 - 4 | 387 | 88 | ||
XM_017019879.1 | XP_016875368.1 | 5 - 6 | 776 | 88 | ||
XM_017019880.1 | XP_016875369.1 | 3 - 4 | 387 | 88 | ||
XM_017019881.1 | XP_016875370.1 | 5 - 6 | 776 | 88 | ||
XM_017019882.1 | XP_016875371.1 | 5 - 6 | 776 | 88 | ||
XM_017019883.1 | XP_016875372.1 | 3 - 4 | 387 | 88 | ||
GenBank mRNA | AK021453.1 | - | 4 - 5 | 408 | 88 | |
AY074491.1 | - | 4 - 5 | 1558 | 88 | ||
BX537569.1 | - | 4 - 5 | 707 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Homo sapiens 1700 Probe spans exons |