Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0200174577.hg.1 |
ENST00000324106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174577.hg.1 |
ENST00000374481.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174577.hg.1 |
ENST00000458610.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174577.hg.1 |
NM_001243077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174577.hg.1 |
NM_001243078.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174577.hg.1 |
NM_006139.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174577.hg.1 |
ENST00000324106.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174577.hg.1 |
ENST00000374481.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174577.hg.1 |
ENST00000458610.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200174577.hg.1 |
NM_001243077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174577.hg.1 |
NM_001243078.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174577.hg.1 |
NM_006139.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200174577.hg.1 |
TC0200010523.hg.1 |
Clariom D TC |