Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028276.hg.1 |
TC07001823.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028276.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028276.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028275.hg.1 |
TC07001823.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028275.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028275.hg.1 |
ENST00000536685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028275.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028279.hg.1 |
TC07001823.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028279.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028279.hg.1 |
ENST00000536685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028279.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028280.hg.1 |
TC07001823.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028280.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028280.hg.1 |
ENST00000536685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028280.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028273.hg.1 |
TC07001823.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028273.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028273.hg.1 |
ENST00000536685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028273.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028277.hg.1 |
TC07001823.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028277.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028277.hg.1 |
ENST00000536685.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028277.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028282.hg.1 |
TC07001823.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028282.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07028282.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189831.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189831.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189831.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189831.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189831.hg.1 |
TC0700012461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700189829.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189829.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189829.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189829.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189829.hg.1 |
TC0700012461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700189830.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189830.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189830.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189830.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189830.hg.1 |
TC0700012461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700189832.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189832.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189832.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189832.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189832.hg.1 |
TC0700012461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700189833.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189833.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189833.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700189833.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700189833.hg.1 |
TC0700012461.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
ENST00000223023.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012461.hg.1 |
NM_003941.3 |
Refseq |