Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010720.hg.1 |
ENST00000419474.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010720.hg.1 |
ENST00000560206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010720.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010720.hg.1 |
TC14000961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010729.hg.1 |
ENST00000382785.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010729.hg.1 |
ENST00000419474.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010729.hg.1 |
ENST00000543520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010729.hg.1 |
ENST00000560206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010729.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010729.hg.1 |
TC14000961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010735.hg.1 |
ENST00000419474.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010735.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010735.hg.1 |
TC14000961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010736.hg.1 |
ENST00000382785.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010736.hg.1 |
ENST00000419474.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010736.hg.1 |
ENST00000560206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010736.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010736.hg.1 |
TC14000961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010725.hg.1 |
ENST00000419474.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010725.hg.1 |
ENST00000560206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010725.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010725.hg.1 |
TC14000961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010734.hg.1 |
ENST00000419474.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010734.hg.1 |
ENST00000543520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010734.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010734.hg.1 |
TC14000961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000606808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000606808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010723.hg.1 |
ENST00000419474.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010723.hg.1 |
ENST00000560206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010723.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010723.hg.1 |
TC14000961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010731.hg.1 |
ENST00000382785.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010731.hg.1 |
ENST00000419474.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010731.hg.1 |
ENST00000543520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010731.hg.1 |
ENST00000560206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010731.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010731.hg.1 |
TC14000961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010737.hg.1 |
ENST00000382785.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010737.hg.1 |
ENST00000419474.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010737.hg.1 |
ENST00000560206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010737.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010737.hg.1 |
TC14000961.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400163539.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163539.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163539.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163539.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163539.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163539.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163539.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400163541.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163541.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163541.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163541.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163541.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163541.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163541.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400163542.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163542.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163542.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163542.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163542.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400163543.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163543.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163543.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163543.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163543.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400163544.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163544.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163544.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163544.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163544.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400163545.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163545.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163545.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163545.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163545.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163545.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163545.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000606808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000606808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163534.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163534.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163534.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163534.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163534.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400163537.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163537.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163537.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163537.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163537.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400163540.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163540.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163540.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163540.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163540.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163540.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163540.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400163536.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163536.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163536.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163536.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163536.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |