Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023283.hg.1 |
ENST00000355311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023283.hg.1 |
ENST00000368648.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023283.hg.1 |
ENST00000471715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023283.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023283.hg.1 |
TC10001761.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023281.hg.1 |
ENST00000355311.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023281.hg.1 |
ENST00000368648.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023281.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023281.hg.1 |
TC10001761.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000440978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000440978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000440978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000440978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000440978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000440978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
ENST00000440978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000440978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000440978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000440978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000440978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000440978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000440978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
ENST00000440978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012270.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183766.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183766.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183766.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183766.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183766.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183766.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183766.hg.1 |
TC1000012270.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000183765.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183765.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183765.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183765.hg.1 |
ENST00000355311.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183765.hg.1 |
ENST00000368648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000183765.hg.1 |
NM_001005463.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000183765.hg.1 |
TC1000012270.hg.1 |
Clariom D TC |