Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004434.hg.1 |
ENST00000212015.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004434.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004434.hg.1 |
ENST00000406900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004434.hg.1 |
ENST00000432464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004434.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004434.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004434.hg.1 |
TC10000400.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000473922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000473922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000473922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000473922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000473922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000497639.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000497639.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000497639.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
ENST00000497639.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000473922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000473922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000473922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000473922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000473922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000497639.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000497639.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000497639.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
ENST00000497639.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007846.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153029.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153029.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153029.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153029.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153029.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153029.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153029.hg.1 |
ENST00000212015.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153029.hg.1 |
ENST00000403579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153029.hg.1 |
ENST00000406900.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153029.hg.1 |
ENST00000432464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153029.hg.1 |
NM_001142498.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153029.hg.1 |
NM_012238.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153029.hg.1 |
TC1000007846.hg.1 |
Clariom D TC |