Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306080.1 | NP_001293009.1 | 21 - 22 | 3843 | 113 | |
NM_005358.5 | NP_005349.3 | 20 - 21 | 3887 | 113 | ||
NM_015842.2 | NP_056667.2 | 18 - 19 | 3534 | 113 | ||
XM_011535078.1 | XP_011533380.1 | 21 - 22 | 3689 | 113 | ||
XM_011535082.1 | XP_011533384.1 | 21 - 22 | 3608 | 113 | ||
XM_011535086.1 | XP_011533388.1 | 20 - 21 | 3404 | 113 | ||
XM_011535094.1 | XP_011533396.1 | 15 - 16 | 2312 | 113 | ||
XM_011535095.1 | XP_011533397.1 | 18 - 19 | 2483 | 113 | ||
XM_017020575.1 | XP_016876064.1 | 21 - 22 | 3689 | 113 | ||
XM_017020579.1 | XP_016876068.1 | 21 - 22 | 3608 | 113 | ||
XM_017020587.1 | XP_016876076.1 | 19 - 20 | 3122 | 113 | ||
XM_017020588.1 | XP_016876077.1 | 19 - 20 | 3203 | 113 | ||
XM_017020589.1 | XP_016876078.1 | 19 - 20 | 3326 | 113 | ||
XM_017020590.1 | XP_016876079.1 | 19 - 20 | 3122 | 113 | ||
XM_017020595.1 | XP_016876084.1 | 19 - 20 | 3487 | 113 | ||
XM_017020598.1 | XP_016876087.1 | 19 - 20 | 3487 | 113 | ||
XM_017020601.1 | XP_016876090.1 | 18 - 19 | 3205 | 113 | ||
XM_017020602.1 | XP_016876091.1 | 18 - 19 | 3088 | 113 | ||
XM_017020603.1 | XP_016876092.1 | 18 - 19 | 3205 | 113 | ||
GenBank mRNA | AB020665.2 | - | 18 - 19 | 3464 | 113 | |
AF144237.1 | - | 9 - 10 | 1347 | 113 | ||
AF330045.1 | - | 20 - 21 | 3886 | 113 | ||
AK055465.1 | - | 13 - 14 | 2499 | 113 | ||
AK074265.1 | - | 5 - 6 | 336 | 113 | ||
AK092052.1 | - | 19 - 20 | 2806 | 113 | ||
AK304051.1 | - | 16 - 17 | 2425 | 113 | ||
FJ711162.1 | - | 21 - 22 | 3473 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |