Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027026.hg.1 |
TC06002144.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027026.hg.1 |
ENST00000345567.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027026.hg.1 |
ENST00000354570.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027026.hg.1 |
ENST00000432797.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027026.hg.1 |
ENST00000454590.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027026.hg.1 |
ENST00000544465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027026.hg.1 |
NM_001198608.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027026.hg.1 |
NM_001198609.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027026.hg.1 |
NM_001198614.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027026.hg.1 |
NM_001198615.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027026.hg.1 |
NM_001198617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027026.hg.1 |
NM_001198618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027026.hg.1 |
NM_001198619.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027026.hg.1 |
NM_003980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
ENST00000354570.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
ENST00000432797.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
ENST00000454590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
ENST00000544465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
ENST00000611373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
ENST00000616617.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
ENST00000617204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_001198608.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_001198609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_001198614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_001198615.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_001198617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_001198618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_001198619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_003980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
ENST00000354570.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
ENST00000432797.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
ENST00000454590.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
ENST00000544465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
ENST00000611373.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
ENST00000616617.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
ENST00000617204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_001198608.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_001198609.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_001198614.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_001198615.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_001198617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_001198618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_001198619.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
NM_003980.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191899.hg.1 |
TC0600013275.hg.1 |
Clariom D TC |