Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028865.hg.1 |
TC11003495.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028865.hg.1 |
ENST00000326621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028865.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028865.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100197484.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100197484.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100197484.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100197484.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100197484.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100197484.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100197484.hg.1 |
TC1100012685.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |