Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01051462.hg.1 |
TC01003315.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01051462.hg.1 |
ENST00000368346.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01051462.hg.1 |
ENST00000392403.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01051462.hg.1 |
NM_018489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100220534.hg.1 |
ENST00000368346.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220534.hg.1 |
ENST00000392403.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220534.hg.1 |
ENST00000492987.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220534.hg.1 |
NM_018489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100220534.hg.1 |
ENST00000368346.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220534.hg.1 |
ENST00000392403.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220534.hg.1 |
ENST00000492987.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220534.hg.1 |
NM_018489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100220534.hg.1 |
TC0100015957.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100220535.hg.1 |
ENST00000368346.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220535.hg.1 |
ENST00000392403.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220535.hg.1 |
ENST00000492987.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220535.hg.1 |
NM_018489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100220535.hg.1 |
ENST00000368346.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220535.hg.1 |
ENST00000392403.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220535.hg.1 |
ENST00000492987.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100220535.hg.1 |
NM_018489.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100220535.hg.1 |
TC0100015957.hg.1 |
Clariom D TC |