Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023342.hg.1 |
ENST00000252825.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023342.hg.1 |
ENST00000391863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023342.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023342.hg.1 |
TC19001709.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193122.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193122.hg.1 |
ENST00000595625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193122.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193122.hg.1 |
TC1900011127.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193122.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193122.hg.1 |
ENST00000595625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193122.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000252825.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595167.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000595625.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
ENST00000598858.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011127.hg.1 |
NM_002152.2 |
Refseq |