Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019701.hg.1 |
TC02001292.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019701.hg.1 |
ENST00000318507.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019701.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019701.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019696.hg.1 |
TC02001292.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019696.hg.1 |
ENST00000318507.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019696.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000415392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000415392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000415392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000415392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000415392.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000418878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000418878.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000428565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000428565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000428565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000428565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000428565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000428565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000449014.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000449014.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000449014.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000449014.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019698.hg.1 |
TC02001292.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019698.hg.1 |
ENST00000318507.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019698.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019699.hg.1 |
TC02001292.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019699.hg.1 |
ENST00000318507.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019699.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019699.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019688.hg.1 |
TC02001292.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019688.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
ENST00000415392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
ENST00000428565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
ENST00000449014.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
ENST00000415392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
ENST00000428565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
ENST00000449014.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176534.hg.1 |
TC0200010785.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000415392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000415392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000415392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000415392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000415392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000418878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000418878.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000428565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000428565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000428565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000428565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000428565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000428565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000449014.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000449014.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000449014.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000449014.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010785.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176528.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176528.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176528.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176528.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176528.hg.1 |
TC0200010785.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
ENST00000415392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
ENST00000449014.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
ENST00000415392.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
ENST00000449014.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176530.hg.1 |
TC0200010785.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176517.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176517.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176517.hg.1 |
ENST00000453237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176517.hg.1 |
NM_001168298.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176517.hg.1 |
TC0200010785.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176526.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176526.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176526.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176526.hg.1 |
ENST00000318507.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176526.hg.1 |
ENST00000454148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176526.hg.1 |
NM_001557.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176526.hg.1 |
TC0200010785.hg.1 |
Clariom D TC |