Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000012049.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000366382.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000591606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000591606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000591606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000591606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000591606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000592769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000592769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000592769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000592769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
ENST00000592769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_001163377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008343.hg.1 |
NM_017659.3 |
Refseq |