Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252049.1 | NP_001238978.1 | 4 - 5 | 500 | 127 | |
NM_001252051.1 | NP_001238980.1 | 3 - 4 | 395 | 127 | ||
NM_001252053.1 | NP_001238982.1 | 4 - 5 | 500 | 127 | ||
NM_005389.2 | NP_005380.2 | 4 - 5 | 500 | 127 | ||
XM_011535868.2 | XP_011534170.1 | 4 - 5 | 701 | 127 | ||
GenBank mRNA | AK129778.1 | - | 4 - 5 | 416 | 127 | |
AK289724.1 | - | 4 - 5 | 468 | 127 | ||
BC007501.2 | - | 4 - 5 | 442 | 3010742 | 127 | |
BC008748.2 | - | 4 - 5 | 378 | 3346722 | 127 | |
CX786396.1 | - | 4 - 5 | 386 | 127 | ||
D13892.1 | - | 4 - 5 | 443 | 127 | ||
D25545.1 | - | 4 - 5 | 366 | 127 | ||
D25546.1 | - | 4 - 5 | 366 | 127 | ||
D25547.1 | - | 4 - 5 | 366 | 127 | ||
DC356391.1 | - | 4 - 5 | 483 | 127 | ||
DC421281.1 | - | 4 - 5 | 490 | 127 | ||
M93008.1 | - | 4 - 5 | 330 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |