Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000524836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000524836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000524836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000524836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000524836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000525786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000525786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000525786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000530106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000530106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000530106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000530106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000530106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000530203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000530203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000530203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000530203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000538597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000538597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000538597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000545264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000545264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000545264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
ENST00000545264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026505.hg.1 |
ENST00000278520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026505.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026505.hg.1 |
ENST00000542662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026505.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026505.hg.1 |
TC11002212.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100192792.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192792.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192792.hg.1 |
ENST00000545264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192792.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192792.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192792.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192792.hg.1 |
ENST00000545264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192792.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192792.hg.1 |
TC1100012052.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000278520.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000423339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000524836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000524836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000524836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000524836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000524836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000525786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000525786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000525786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000530106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000530106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000530106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000530106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000530106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000530203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000530203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000530203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000530203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000538597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000538597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000538597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000545264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000545264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000545264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
ENST00000545264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012052.hg.1 |
NM_024725.3 |
Refseq |