Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000436333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000436333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000436333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000436333.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000475280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000475280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
ENST00000475280.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000041.hg.1 |
ENST00000397817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000041.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000041.hg.1 |
TC16000004.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000050.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000050.hg.1 |
ENST00000356432.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000050.hg.1 |
ENST00000397817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000050.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000050.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000050.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000050.hg.1 |
TC16000004.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
ENST00000436333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
ENST00000436333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144114.hg.1 |
TC1600006439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000219431.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000356432.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000436333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000436333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000436333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000436333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000475280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000475280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
ENST00000475280.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_001015052.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006439.hg.1 |
NM_002434.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144102.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144102.hg.1 |
ENST00000436333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144102.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144102.hg.1 |
ENST00000397817.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144102.hg.1 |
ENST00000436333.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144102.hg.1 |
NM_001015054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144102.hg.1 |
TC1600006439.hg.1 |
Clariom D TC |