Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017142.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017142.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017142.hg.1 |
TC10001356.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017141.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017141.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017141.hg.1 |
TC10001356.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017143.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017143.hg.1 |
TC10001356.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373307.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373307.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373307.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373307.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373307.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373307.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000373307.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017140.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017140.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017140.hg.1 |
TC10001356.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000174122.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174122.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174122.hg.1 |
TC1000010878.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000174117.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174117.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174117.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174117.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174117.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174117.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174117.hg.1 |
TC1000010878.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000174120.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174120.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174120.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174120.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174120.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174120.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174120.hg.1 |
TC1000010878.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000373307.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010878.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174118.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174118.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174118.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174118.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174118.hg.1 |
ENST00000619173.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174118.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174118.hg.1 |
TC1000010878.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000174121.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174121.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174121.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174121.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174121.hg.1 |
TC1000010878.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000174119.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174119.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174119.hg.1 |
ENST00000373306.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174119.hg.1 |
NM_001057.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174119.hg.1 |
TC1000010878.hg.1 |
Clariom D TC |