Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014159.6 | NP_054878.5 | 7 - 8 | 4977 | 91 | |
XM_011533631.2 | XP_011531933.1 | 8 - 9 | 6473 | 91 | ||
XM_011533632.2 | XP_011531934.1 | 7 - 8 | 5049 | 91 | ||
XM_011533633.2 | XP_011531935.1 | 8 - 9 | 6475 | 91 | ||
XM_011533634.1 | XP_011531936.1 | 7 - 8 | 5002 | 91 | ||
GenBank mRNA | AB051519.2 | - | 5 - 6 | 2977 | 91 | |
AF161554.1 | - | 5 - 6 | 721 | 91 | ||
AJ238403.1 | - | 5 - 6 | 3824 | 91 | ||
AL831959.1 | - | 4 - 5 | 3952 | 91 | ||
AY576987.1 | - | 5 - 6 | 3486 | 91 | ||
AY576988.1 | - | 5 - 6 | 3486 | 91 | ||
BC090954.1 | - | 5 - 6 | 2767 | 91 | ||
BC117162.1 | - | 5 - 6 | 3600 | 40125713 | 91 | |
BC117164.1 | - | 5 - 6 | 3600 | 40125715 | 91 | |
BX649110.1 | - | 5 - 6 | 4744 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |