Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282613.1 | NP_001269542.1 | 1 - 2 | 202 | 112 | |
NM_001282615.1 | NP_001269544.1 | 1 - 2 | 202 | 112 | ||
GenBank mRNA | BC006440.1 | - | 1 - 2 | 119 | 112 | |
BC151138.1 | - | 1 - 2 | 71 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons |