Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLSTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014718.3 | NP_055533.2 | 16 - 17 | 2801 | 127 | |
XM_006719163.3 | XP_006719226.1 | 15 - 16 | 4127 | 127 | ||
GenBank mRNA | AB018269.1 | - | 15 - 16 | 3097 | 127 | |
AJ277460.1 | - | 16 - 17 | 2798 | 127 | ||
AK299177.1 | - | 16 - 17 | 2775 | 127 | ||
AK299246.1 | - | 7 - 8 | 1003 | 127 | ||
AK316059.1 | - | 16 - 17 | 2801 | 127 | ||
AY753302.1 | - | 15 - 16 | 2559 | 127 | ||
BC104767.1 | - | 16 - 17 | 2605 | 8143770 | 127 | |
BC111491.1 | - | 16 - 17 | 2789 | 5213830 | 127 | |
BC112283.1 | - | 16 - 17 | 2617 | 8327751 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |