Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003000.hg.1 |
ENST00000316843.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003000.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003000.hg.1 |
TC17000208.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003001.hg.1 |
ENST00000316843.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003001.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003001.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17003001.hg.1 |
TC17000208.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700150209.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700150209.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700150209.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700150209.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700150209.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700150209.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700150209.hg.1 |
TC1700007105.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007105.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700150210.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700150210.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700150210.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700150210.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700150210.hg.1 |
ENST00000316843.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700150210.hg.1 |
ENST00000479155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700150210.hg.1 |
ENST00000621229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700150210.hg.1 |
NM_004140.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700150210.hg.1 |
TC1700007105.hg.1 |
Clariom D TC |