Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000571079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000571079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000571079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000571079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000571597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000571597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000571597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000571597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000571597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000571597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000571597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000573057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000573057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000573057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000573057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000573957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000573957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000573957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000573957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000574490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000574490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000574490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000576693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000576693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000576693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000576693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
ENST00000576693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014673.hg.1 |
ENST00000250113.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014673.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014673.hg.1 |
TC17001092.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700174924.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174924.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700174924.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700174924.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700174924.hg.1 |
TC1700009649.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000250113.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000571079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000571079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000571079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000571079.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000571597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000571597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000571597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000571597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000571597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000571597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000571597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000573057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000573057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000573057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000573057.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000573957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000573957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000573957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000573957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000574490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000574490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000574490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000576693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000576693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000576693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000576693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
ENST00000576693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009649.hg.1 |
NM_004860.3 |
Refseq |