Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS26A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318946.1 | NP_001305875.1 | 2 - 3 | 284 | 158 | |
GenBank mRNA | AK290536.1 | - | 2 - 3 | 190 | 158 | |
BX647725.1 | - | 2 - 3 | 205 | 158 | ||
BX649092.1 | - | 2 - 3 | 259 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |