Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372.3 | NP_001363.2 | 30 - 31 | 6172 | 68 | |
XM_011523703.1 | XP_011522005.1 | 30 - 31 | 6136 | 68 | ||
XM_017024292.1 | XP_016879781.1 | 29 - 30 | 6019 | 68 | ||
XM_017024293.1 | XP_016879782.1 | 19 - 20 | 4080 | 68 | ||
XM_017024294.1 | XP_016879783.1 | 30 - 31 | 6136 | 68 | ||
XM_017024295.1 | XP_016879784.1 | 30 - 31 | 6136 | 68 | ||
GenBank mRNA | AB002355.2 | - | 11 - 12 | 1850 | 68 | |
AB290158.1 | - | 18 - 19 | 3989 | 68 | ||
AF257737.1 | - | 30 - 31 | 6162 | 68 | ||
AJ404468.1 | - | 30 - 31 | 6172 | 68 | ||
BC153883.1 | - | 11 - 12 | 1850 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |