Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17002153.hg.1 |
ENST00000262442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17002153.hg.1 |
ENST00000454412.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17002153.hg.1 |
NM_001372.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17002153.hg.1 |
TC17000154.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700148601.hg.1 |
ENST00000262442.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148601.hg.1 |
ENST00000454412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148601.hg.1 |
NM_001372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700148601.hg.1 |
ENST00000262442.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148601.hg.1 |
ENST00000454412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148601.hg.1 |
NM_001372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700148601.hg.1 |
TC1700006897.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700148600.hg.1 |
ENST00000262442.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148600.hg.1 |
ENST00000454412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148600.hg.1 |
ENST00000579703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148600.hg.1 |
NM_001372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700148600.hg.1 |
ENST00000262442.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148600.hg.1 |
ENST00000454412.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148600.hg.1 |
ENST00000579703.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148600.hg.1 |
NM_001372.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700148600.hg.1 |
TC1700006897.hg.1 |
Clariom D TC |