Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015945.hg.1 |
ENST00000264833.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015945.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015945.hg.1 |
TC19001155.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000590410.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000590410.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000590410.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000590410.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000590410.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177410.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177410.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177410.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177410.hg.1 |
TC1900009603.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900177410.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177410.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177410.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
ENST00000590410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
TC1900009603.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
ENST00000590410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177408.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
ENST00000590410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
TC1900009603.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
ENST00000590410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900177407.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000264833.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000590410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000590410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000590410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000590410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000590410.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000590841.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000592448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
ENST00000593091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304347.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_001304348.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009603.hg.1 |
NM_058164.3 |
Refseq |