Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000553535.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000553535.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000553535.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000553535.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000553535.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000554986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000554986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000554986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000554986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000554986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000554986.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000555627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000555627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000555627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000555627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
NM_005982.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
NM_005982.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
NM_005982.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
NM_005982.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
NM_005982.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
NM_005982.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009351.hg.1 |
NM_005982.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000247182.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000553535.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000553535.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000553535.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000553535.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000553535.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000554986.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000554986.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000554986.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000554986.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000554986.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000554986.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000555627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000555627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000555627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000555627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
ENST00000555955.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
NM_005982.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
NM_005982.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
NM_005982.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
NM_005982.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
NM_005982.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
NM_005982.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009351.hg.1 |
NM_005982.3 |
Refseq |