Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000462175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000462175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000462175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000470325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000470325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000470325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000470325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000470325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000470325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000536146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000536146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000536146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000536146.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000539316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000539316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000539316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000539316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000539316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000543896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000543896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000543896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000543896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000543896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000578011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000578011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000578011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
ENST00000578011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015688.hg.1 |
ENST00000338696.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015688.hg.1 |
ENST00000395930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015688.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015688.hg.1 |
TC17001162.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700176842.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700176842.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700176842.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700176842.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700176842.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700176842.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700176842.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700176842.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700176842.hg.1 |
TC1700009857.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000338696.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000395931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000462175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000462175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000462175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000470325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000470325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000470325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000470325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000470325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000470325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000536146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000536146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000536146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000536146.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000539245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000539316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000539316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000539316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000539316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000539316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000543896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000543896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000543896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000543896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000543896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000578011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000578011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000578011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
ENST00000578011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009857.hg.1 |
NM_031898.2 |
Refseq |