Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000505529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000505529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000505529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000505529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000505529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000511199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000511199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000511199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000511199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000511199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
ENST00000511199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001743.hg.1 |
TC04000080.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001743.hg.1 |
ENST00000329016.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001743.hg.1 |
ENST00000507866.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001743.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146603.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146603.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146603.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146603.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146603.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400146603.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400146603.hg.1 |
TC0400006718.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000329016.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000505529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000505529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000505529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000505529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000505529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000507866.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000511199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000511199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000511199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000511199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000511199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
ENST00000511199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006718.hg.1 |
NM_020777.2 |
Refseq |